Thermo Scientific Chemicals

Calcium iodide hydrate, Thermo Scientific Chemicals

Catalog number: 040136.36
500 g, Each
Thermo Scientific Chemicals

Calcium iodide hydrate, Thermo Scientific Chemicals

Catalog number: 040136.36
500 g, Each
Quantity
Catalog number: 040136.36
also known as 040136-36
Price (USD)
Price: 393.00
Online price: 334.65
Your price:
Quantity
-

Chemical Identifiers

CAS
71626-98-7
IUPAC Name
calcium diiodide
Molecular Formula
CaI2
InChI Key
UNMYWSMUMWPJLR-UHFFFAOYSA-L
SMILES
[Ca++].[I-].[I-]
Assay (unspecified)
>=97.5%

Description

Calcium iodide finds use in the preparation of complex used in polymerization and hydroamination as well as serving as a matrix for luminescent materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text