Thermo Scientific Chemicals

Magnesium silicate monohydrate (Talc), Thermo Scientific Chemicals

Catalog number: 040318.22
100 g, Each
Thermo Scientific Chemicals

Magnesium silicate monohydrate (Talc), Thermo Scientific Chemicals

Catalog number: 040318.22
100 g, Each
Quantity
Catalog number: 040318.22
also known as 040318-22
Price (USD)
Price: 27.50
Online price: 23.65
Your price:
Quantity
-

Chemical Identifiers

CAS
14807-96-6
IUPAC Name
trimagnesium(2+) bis(oxosilanebis(olate)) dihydroxysilanoylolate
Molecular Formula
H2Mg3O12Si4
InChI Key
KNXVOGGZOFOROK-UHFFFAOYSA-N
SMILES
[Mg++].[Mg++].[Mg++].O[Si]([O-])=O.O[Si]([O-])=O.[O-][Si]([O-])=O.[O-][Si]([O-])=O
Assay (unspecified)
17-19.5% Mg

Description

Magnesium silicate is used as a lubricant, filler, electric and heat insulator and in clarifying liquids by filtration. Talc is used in many industries such as paper making, plastic, paints and coatings, rubber, food, electric cable, pharmaceuticals, cosmetics and ceramics. It is a widely used as baby powder (aka talcum). It is also useful for the treatment of complicated spontaneous pneumothorax.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text