Gold(I)-chlorid, Premion™, 99.99 % (Metallbasis), Au 84.2 % min., Thermo Scientific Chemicals
Gold(I)-chlorid, Premion™, 99.99 % (Metallbasis), Au 84.2 % min., Thermo Scientific Chemicals
Gold(I)-chlorid, Premion™, 99.99 % (Metallbasis), Au 84.2 % min., Thermo Scientific Chemicals
Thermo Scientific Chemicals

Gold(I)-chlorid, Premion™, 99.99 % (Metallbasis), Au 84.2 % min., Thermo Scientific Chemicals

CAS: 10294-29-8 | AuCl | 232.42 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
0.25 g
1 g
5 g
Katalognummer 040432.03
auch als 040432-03 bezeichnet
Preis (EUR)
275,00
Each
Menge:
1 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
275,00
Each
Chemikalien-Kennzeichnungen
CAS10294-29-8
IUPAC Namegold(1+) chloride
Molecular FormulaAuCl
InChI KeyFDWREHZXQUYJFJ-UHFFFAOYSA-M
SMILES[Cl-].[Au+]
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Assay (unspecified)Au 84.2% min
Total Metal Impurities0.01% max.
As intermediates. Also In Chemicals, pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only