Thermo Scientific Chemicals

Niobium powder, -325 mesh, Puratronic™, 99.99% (metals basis excluding Ta), Ta |<500ppm, Thermo Scientific Chemicals

Catalog number: 040510.18
50 g, Each
Thermo Scientific Chemicals

Niobium powder, -325 mesh, Puratronic™, 99.99% (metals basis excluding Ta), Ta |<500ppm, Thermo Scientific Chemicals

Catalog number: 040510.18
50 g, Each
Quantity
Catalog number: 040510.18
also known as 040510-18
Price (USD)

Chemical Identifiers

CAS
7440-03-1
IUPAC Name
niobium
Molecular Formula
Nb
InChI Key
GUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES
[Nb]
Appearance (Color)
Grey to dark grey
Form
Powder or crystals
Total Metal Impurities excluding Ta
≤0.01% max.
Particle Size
-325 mesh ( less than 45 micron )
Trace Metal
Total metal impurities less than 0.01% excluding Ta, Ta <500ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text