Polyethylenimin, verzweigt, Molekulargewicht 600, 99 %, Thermo Scientific Chemicals
Polyethylenimin, verzweigt, Molekulargewicht 600, 99 %, Thermo Scientific Chemicals
Polyethylenimin, verzweigt, Molekulargewicht 600, 99 %, Thermo Scientific Chemicals
Polyethylenimin, verzweigt, Molekulargewicht 600, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polyethylenimin, verzweigt, Molekulargewicht 600, 99 %, Thermo Scientific Chemicals

CAS: 9002-98-6 | ((C2H5N)zC2H4N)y(C2H5N)x | 43.07 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
25 g
100 g
500 g
Katalognummer 040527.22
auch als 040527-22 bezeichnet
Preis (EUR)
214,00
Each
Menge:
100 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
214,00
Each
Chemikalien-Kennzeichnungen
CAS9002-98-6
Molecular Formula((C2H5N)zC2H4N)y(C2H5N)x
SMILES*-CCNCCN(-*)CCN-*
MDL NumberMFCD00084427
SpecificationsSpecification SheetDatenblatt
Appearance (Color)Light yellow
Formclear viscous liquid
Assay (unspecified)99.0%, min
Polyethyleneimine, branched, M.W. 600 acts as an auxiliary agent. It is used in paper-making, water treatment, detergents, adhesives and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only