Hafnium(IV)-oxid, 99.99 % (Metallbasis ohne Zr), Zr < 100 ppm, Thermo Scientific Chemicals
Hafnium(IV)-oxid, 99.99 % (Metallbasis ohne Zr), Zr < 100 ppm, Thermo Scientific Chemicals
Hafnium(IV)-oxid, 99.99 % (Metallbasis ohne Zr), Zr < 100 ppm, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hafnium(IV)-oxid, 99.99 % (Metallbasis ohne Zr), Zr < 100 ppm, Thermo Scientific Chemicals

Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
1 g
5 g
Katalognummer 040620.03
Preis (EUR)
-
Menge:
1 g
Großbestellung oder individuelle Größe anfordern
Chemikalien-Kennzeichnungen
CAS12055-23-1
IUPAC Namehafnium(4+) dioxidandiide
Molecular FormulaHfO2
InChI KeyWIHZLLGSGQNAGK-UHFFFAOYSA-N
SMILES[O--].[O--].[Hf+4]
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Impurity contentZr <100ppm
Total Metal Impurities excluding Zr0.01% max.
Hafnium(IV) oxide is used as Intermediates, Paint additives and coating additives, metal Products. And it is also used in optical coatings, as a refractory material in the insulation of such devices as thermocouples.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only