Thermo Scientific Chemicals

Lámina ultrafina de aluminio, 0,8 micrómetros de grosor, 99,1% (base metálica), Thermo Scientific Chemicals

Número de catálogo: 040707.YD
30 x 30 mm, Each
Thermo Scientific Chemicals

Lámina ultrafina de aluminio, 0,8 micrómetros de grosor, 99,1% (base metálica), Thermo Scientific Chemicals

Número de catálogo: 040707.YD
30 x 30 mm, Each
Cantidad
Número de catálogo: 040707.YD
también denominado 040707-YD
Precio (EUR)
Cantidad
-

Identificadores químicos

CAS
7429-90-5
IUPAC Name
aluminium
Molecular Formula
Al
InChI Key
XAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES
[Al]
Appearance (Color)
Silver
Form
Foil, 0.8 micron thick.
Assay from Supplier's CofA
≥99.1% (metals basis)

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO - Para uso exclusivo en investigación

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text