Edelstahl-Gaze, 20 Mesh gewebet aus Draht mit Durchmesser von 0.382 mm (0.015), Typ 304, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Edelstahl-Gaze, 20 Mesh gewebet aus Draht mit Durchmesser von 0.382 mm (0.015), Typ 304, Thermo Scientific Chemicals

Filtration, Elektroden
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
75 x 75 mm
150 x 150 mm
300 x 300 mm
Katalognummer 040942.FL
auch als 040942-FL bezeichnet
Preis (EUR)
37,00
Each
Menge:
75 x 75 mm
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
37,00
Each
Specifications
Durchschnitt Mol. Gew. oder Mol.-Gew. GewichtsbereichFe:Cr:Ni; 70:19:11 wt%
Chemischer Name oder MaterialStainless Steel gauze, Type 304
MDL-NummerMFCD00148499
Netzgröße20 mesh
NamenshinweisWoven from 0.382mm (0.015 in.) dia. wire
Mehr anzeigen
Stainless Steel gauze, Type 316 is used to woven wire cloth which has extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only