Thermo Scientific Chemicals

Titanium nitride, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 041556.A1
1 kg, Each
Thermo Scientific Chemicals

Titanium nitride, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog number: 041556.A1
1 kg, Each
Quantity
Catalog number: 041556.A1
also known as 041556-A1
Price (USD)
Price: 902.00
Online price: 780.65
Your price:
Quantity
-

Chemical Identifiers

CAS
25583-20-4
IUPAC Name
titanioamine
Molecular Formula
NTi
InChI Key
NRTOMJZYCJJWKI-UHFFFAOYSA-N
SMILES
N#[Ti]
Total Metal Impurities
0.5% max.

Description

Titanium nitride is used as a processing agent in powder metallurgy and laboratory. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff. It is used as Use as primary and secondary intermediate. Artificial limbs; Barrier layer in contact and interconnect metallization; Biological materials cutting tools; Gate electrode in metal-oxide-semiconductor (MOS) transistors; Low-barrier Schottky diode; Optical devices in aggressive environments; Plastic molds; Prostheses; Wear-resistant coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text