Antimon(III)-bromid, 99 %, Thermo Scientific Chemicals
Antimon(III)-bromid, 99 %, Thermo Scientific Chemicals
Antimon(III)-bromid, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Antimon(III)-bromid, 99 %, Thermo Scientific Chemicals

CAS: 7789-61-9 | Br3Sb | 361.472 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
50 g
250 g
Katalognummer 041573.30
auch als 041573-30 bezeichnet
Preis (EUR)
756,00
Each
Menge:
250 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
756,00
Each
Chemikalien-Kennzeichnungen
CAS7789-61-9
SpecificationsSpecification SheetDatenblatt
Assay (unspecified)98.6% min
Antimony(III) bromide is used in the synthesis of antimony compounds, in semiconductors and chemical analysis. It acts as a fire retardant in polyethylene polymers, as mordant and in dyeing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only