N,N-Dimethylformamid, HPLC-Gütegrad, >99.7 %, Thermo Scientific Chemicals
N,N-Dimethylformamid, HPLC-Gütegrad, >99.7 %, Thermo Scientific Chemicals
N,N-Dimethylformamid, HPLC-Gütegrad, >99.7 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N-Dimethylformamid, HPLC-Gütegrad, >99.7 %, Thermo Scientific Chemicals

Die gleichen HPLC-Spezifikationen wie 22915
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
250 mL
1 L
Katalognummer 041859.AK
auch als 041859-AK bezeichnet
Preis (EUR)
33,20
Each
Menge:
250 mL
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
33,20
Each
Chemikalien-Kennzeichnungen
CAS68-12-2
IUPAC NameN,N-dimethylformamide
Molecular FormulaC3H7NO
InChI KeyZMXDDKWLCZADIW-UHFFFAOYSA-N
SMILESCN(C)C=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)Colorless
UV/VisAt 270 nm 1.00 AU, max
UV/VisAt 295 nm 0.10 AU, max
UV/VisAt 340 nm 0.01 AU, max
UV/VisAt 275 nm 0.30 AU, max
Mehr anzeigen
N,N-Dimethylformamide is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only