Thermo Scientific Chemicals

Ethylene glycol diacetate, 97%, Thermo Scientific Chemicals

Catalog number: 041910.22
100 g, Each
Thermo Scientific Chemicals

Ethylene glycol diacetate, 97%, Thermo Scientific Chemicals

Catalog number: 041910.22
100 g, Each
Quantity
Catalog number: 041910.22
also known as 041910-22
Price (USD)
Price: 18.20
Online price: 14.65
Your price:
Quantity
-

Chemical Identifiers

CAS
111-55-7
Appearance (Color)
Colorless
Assay (GC)
≥96.0%
Water Content (Karl Fischer Titration)
≤0.2%
Refractive Index
1.412 to 1.416
Form
Liquid

Description

EGDA imparts excellent flow properties in baking lacquers and enamels and where thermoplastic acrylic resins are used. It is also a good solvent for cellulosic coatings and can be used in some ink systems such as screen inks. It has found use as a perfume fixative, and has reported applications in waterborne adhesives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text