Aluminiumacrylat, Thermo Scientific Chemicals
Aluminiumacrylat, Thermo Scientific Chemicals
Aluminiumacrylat, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminiumacrylat, Thermo Scientific Chemicals

CAS: 15743-20-1 | C9H12AlO6 | 243.171 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
5 g
25 g
100 g
Katalognummer 042003.14
auch als 042003-14 bezeichnet
Preis (EUR)
179,00
Each
Menge:
25 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
179,00
Each
Specifications
CAS15743-20-1
Chemischer Name oder MaterialAluminum acrylate
EINECS-Nummer239-836-8
Formelmasse243.17
Gesundheitsgefahr 1Gehäuse H315-H319-H335
Mehr anzeigen
Aluminum acrylate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only