Sechswertiges Chrom, Standardlösung, Specpure™, Cr+6 1000 μg/ml, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sechswertiges Chrom, Standardlösung, Specpure™, Cr+6 1000 μg/ml, Thermo Scientific Chemicals

Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
100 mL
500 mL
Katalognummer 042234.AE
auch als 042234-AE bezeichnet
Preis (EUR)
116,65
Exklusiv online
129,00
Ersparnis 12,35 (10%)
Each
Menge:
100 mL
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
116,65
Exklusiv online
129,00
Ersparnis 12,35 (10%)
Each
Specifications
Chemischer Name oder MaterialHexavalent Chromium standard solution
FarbeGelb
EINECS-Nummer232-143-1
Gesundheitsgefahr 1Gehäuse H340-H350-H360
Gesundheitsgefahr 2GHS H Statement
H334-H340-H350-H360-H317-H208
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
May cause an allergic skin reaction.
Mehr anzeigen
Hexavalent Chromium, standard solution is used in ion chromatography for measurement and determination of soluble Cr(VI).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only