Mangan(IV)-oxid, 99.9 % (Metallbasis), Thermo Scientific Chemicals
Mangan(IV)-oxid, 99.9 % (Metallbasis), Thermo Scientific Chemicals
Mangan(IV)-oxid, 99.9 % (Metallbasis), Thermo Scientific Chemicals
Mangan(IV)-oxid, 99.9 % (Metallbasis), Thermo Scientific Chemicals
Mangan(IV)-oxid, 99.9 % (Metallbasis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Mangan(IV)-oxid, 99.9 % (Metallbasis), Thermo Scientific Chemicals

CAS: 1313-13-9 | MnO2 | 86.94 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
250 g
1 kg
Katalognummer 042250.30
auch als 042250-30 bezeichnet
Preis (EUR)
110,00
Each
Zum Warenkorb hinzufügen
Menge:
250 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
110,00
Each
Zum Warenkorb hinzufügen
Chemikalien-Kennzeichnungen
CAS1313-13-9
IUPAC Namedioxomanganese
Molecular FormulaMnO2
InChI KeyNUJOXMJBOLGQSY-UHFFFAOYSA-N
SMILESO=[Mn]=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Total Metal Impurities0.1% max.
It is used as an oxidizing agent in organic synthesis such as oxidation of allylic/benzylic alcohols, as a textile dye, as a reducing agent, and as a component of dry cell batteries such as zinc-carbon battery and alkaline battery. It is also used in making pigments for glasses and ceramics, and drier for paints. Further it is used in the manufacture of manganese steel and several manganese derivatives including potassium permanganate, a powerful versatile oxidant. Catalytic hydration of nitriles to amides by flowing through manganese dioxide has been reported. Manganese dioxide supported on inorganic oxide can be used for oxidation of methylamine through CWAO (Catalytic Wet Air Oxidation). It has high potential as highly efficient and robust material for water oxidation reaction (WORs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only