N,O-Bis(trimethylsilyl)trifluoracetamid, ≥ 99 %, verpackt unter Argon in wiederverschließbaren ChemSeal™-Flaschen, Thermo Scientific Chemicals
N,O-Bis(trimethylsilyl)trifluoracetamid, ≥ 99 %, verpackt unter Argon in wiederverschließbaren ChemSeal™-Flaschen, Thermo Scientific Chemicals
N,O-Bis(trimethylsilyl)trifluoracetamid, ≥ 99 %, verpackt unter Argon in wiederverschließbaren ChemSeal™-Flaschen, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,O-Bis(trimethylsilyl)trifluoracetamid, ≥ 99 %, verpackt unter Argon in wiederverschließbaren ChemSeal™-Flaschen, Thermo Scientific Chemicals

CAS: 25561-30-2 | C8H18F3NOSi2 | 257.403 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
5 g
25 g
100 g
Katalognummer 042429.22
auch als 042429-22 bezeichnet
Preis (EUR)
470,00
Each
Menge:
100 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
470,00
Each
Chemikalien-Kennzeichnungen
CAS25561-30-2
IUPAC Nametrimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)ethanimidate
Molecular FormulaC8H18F3NOSi2
InChI KeyXCOBLONWWXQEBS-UHFFFAOYSA-N
SMILESC[Si](C)(C)OC(=N[Si](C)(C)C)C(F)(F)F
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)Clear colorless to very pale yellow
FormLiquid
Assay (GC)99.0%
N,O-Bis(trimethylsilyl)trifluoroacetamide is used as a derivatization reagent for gas chromatography and mass-spectrometry analysis of phenolic acids in fruits. It is also used as a reactant in the preparation of pyrimidinone ribosides and analogs which shows anti-tumor properties. It plays as an important role as a reagent for the regioselective desulfation of polysaccharide sulfates. Furthermore, it is used to prepare trimethylsilyl ethers of carbohydrates and alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Verpackt unter Argon
RUO – Research Use Only