Thermo Scientific Chemicals

Pyrite, naturally occurring mineral, grains, approximately 1.5-4.8mm (0.06-0.19in), Thermo Scientific Chemicals

Catalog number: 042633.14
25 g, Each
Thermo Scientific Chemicals

Pyrite, naturally occurring mineral, grains, approximately 1.5-4.8mm (0.06-0.19in), Thermo Scientific Chemicals

Catalog number: 042633.14
25 g, Each
Quantity
Catalog number: 042633.14
also known as 042633-14
Price (USD)
Price: 396.00
Online price: 336.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1309-36-0
IUPAC Name
disulfanylideneiron
Molecular Formula
FeS2
InChI Key
NFMAZVUSKIJEIH-UHFFFAOYSA-N
SMILES
S=[Fe]=S
Comment
Pyrite is naturally occurring Iron sulfide mined and crushed to approximately 1.5-4.8mm (0.06-0.19in) without any chemical processing.

Description

For grain mounts, XRD and microprobe standards, identification of unknown minerals.Pyrite is used in the preparation of iron(II) sulfate, sulfur dioxide and iron(II) sulfide. It finds application as semiconductor material, as the cathode material in energizer band and non-rechargeable lithium batteries. It is utilized to make marcasite jewelry. In addition, it is used in photovoltaic solar panels. It is useful for grain mounts, XRD and microprobe standards and identification of unknown minerals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Naturally occurring mineral; Hardness: 6-6.5
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text