Thermo Scientific Chemicals

Poly(styrene sulfonic acid), M.W. 75,000, 30% w/v aq. soln., Thermo Scientific Chemicals

Catalog number: 042653.22
100 g, Each
Thermo Scientific Chemicals

Poly(styrene sulfonic acid), M.W. 75,000, 30% w/v aq. soln., Thermo Scientific Chemicals

Catalog number: 042653.22
100 g, Each
Quantity
Catalog number: 042653.22
also known as 042653-22
Price (USD)
Price: 59.70
Online price: 50.65
Your price:
Quantity
-

Chemical Identifiers

CAS
28210-41-5
IUPAC Name
calcium 2-ethenylbenzene-1-sulfonate
Molecular Formula
C8H7CaO3S
InChI Key
IJBYVKWTVVYTPL-UHFFFAOYSA-M
SMILES
[Ca++].[O-]S(=O)(=O)C1=CC=CC=C1C=C
Appearance (Color)
Clear amber
Form
Liquid
Molecular Weight
65000 - 75000
Comment
Solid Content: 29 - 32 %

Description

Poly(styrene sulfonic acid) is used in water softening and to remove hard ions like calcium and magnesium. In the medical field, it is used as potassium binders in acute and chronic kidney disease. Its sodium salt is used as a superplastifier in cement, as a dye improving agent for cotton, as proton exchange membranes in fuel cell applications. It acts as a solid acid catalyst in organic synthesis. It is an antistatic resin used for electrographic and electrophotographic substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text