Tetramethylammoniumformiat, 30 % w/w wässrige Lösung, Thermo Scientific Chemicals
Tetramethylammoniumformiat, 30 % w/w wässrige Lösung, Thermo Scientific Chemicals
Tetramethylammoniumformiat, 30 % w/w wässrige Lösung, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetramethylammoniumformiat, 30 % w/w wässrige Lösung, Thermo Scientific Chemicals

CAS: 59138-84-0 | C5H13NO2 | 119.16 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
100 mL
500 mL
Katalognummer 042710.AP
auch als 042710-AP bezeichnet
Preis (EUR)
309,00
Each
Menge:
500 mL
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
309,00
Each
Chemikalien-Kennzeichnungen
CAS59138-84-0
SpecificationsSpecification SheetDatenblatt
FormLiquid
Assay from Suppliers CofA29.0-31.0%
Tetramethylammonium formate may provide the lowest temperature sensitivity, additionally reducing corrosion of stainless-steel flow system by chloride-induced depassivation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only