Kobalt(II)-isopropoxid, Thermo Scientific Chemicals
Kobalt(II)-isopropoxid, Thermo Scientific Chemicals
Kobalt(II)-isopropoxid, Thermo Scientific Chemicals
Kobalt(II)-isopropoxid, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Kobalt(II)-isopropoxid, Thermo Scientific Chemicals

CAS: 60406-95-3 | C6H14CoO2 | 177.109 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
1 g
5 g
Katalognummer 042810.06
auch als 042810-06 bezeichnet
Preis (EUR)
361,00
Each
Zum Warenkorb hinzufügen
Menge:
5 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
361,00
Each
Zum Warenkorb hinzufügen
Chemikalien-Kennzeichnungen
CAS60406-95-3
SpecificationsSpecification SheetDatenblatt
Appearance (Color)Purple
FormMicrocrystalline powder
Assay from Suppliers CofA≥95.0%

Cobalt(II) isopropoxide is used in display & imaging, catalysis & synthesis and ferroelectric metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampulliert unter Argon
RUO – Research Use Only