Europium(III)-acetathydrat, 99.9 % (REO), Thermo Scientific Chemicals REacton™
Europium(III)-acetathydrat, 99.9 % (REO), Thermo Scientific Chemicals REacton™
Europium(III)-acetathydrat, 99.9 % (REO), Thermo Scientific Chemicals REacton™
Thermo Scientific Chemicals

Europium(III)-acetathydrat, 99.9 % (REO), Thermo Scientific Chemicals REacton™

CAS: 62667-64-5 | C6H9EuO6 | 329.10 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
5 g
25 g
Katalognummer 042878.14
auch als 042878-14 bezeichnet
Preis (EUR)
611,00
Each
Menge:
25 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
611,00
Each
Chemikalien-Kennzeichnungen
CAS62667-64-5
IUPAC Nameeuropium(3+) triacetate
Molecular FormulaC6H9EuO6
InChI KeyLNYNHRRKSYMFHF-UHFFFAOYSA-K
SMILES[Eu+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Total Rare Earth Oxide Impurities0.1% max.
Europium(III) acetate hydrate is used as fluorescent powder and as a chemical reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only