Aceton, Gütegrad, > 99,5 % Thermo Scientific Chemicals
Aceton, Gütegrad, > 99,5 % Thermo Scientific Chemicals
Aceton, Gütegrad, > 99,5 % Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aceton, Gütegrad, > 99,5 % Thermo Scientific Chemicals

Für die Pestizidrückstandsanalyse
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
1 L
4 L
4 x 4 L
Katalognummer 043053.K7
auch als 043053-K7 bezeichnet
Preis (EUR)
120,00
Each
Menge:
4 L
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
120,00
Each
Chemikalien-Kennzeichnungen
CAS67-64-1
IUPAC Namepropan-2-one
Molecular FormulaC3H6O
InChI KeyCSCPPACGZOOCGX-UHFFFAOYSA-N
SMILESCC(C)=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Residue on Evaporation5ppm
Residual halide(as Heptachlorepoxide by GC/ECD) 10ng/L
Residual water0.5%
Suitable for pesticide residue analysisAcetone is a polar, aprotic solvent used in the synthesis and isolation of both organic and inorganic compounds and complexes. It plays a vital role in Jones oxidation. It is utilized for rinsing laboratory glassware due to its low cost and volatility. It is used in the manufacture of various chemicals such as methyl isobutyl ketone, mesityl oxide, diacetone alcohol, acetic acid, iodoform, bromoform, rayon and photographic films, paint and varnish removers. It is also useful in the synthesis of t-butanesulfinamide, terpenes, thiol-stabilized gold colloids, calixphyrin macrocycles and trispyrazolylborate platinum compounds. It plays an important role in protein crystallization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only