Thermo Scientific Chemicals

Silicon(IV) oxide, 30% in H2O, colloidal dispersion, Thermo Scientific Chemicals

產品號碼: 043111.A7
5 kg, Each
Thermo Scientific Chemicals

Silicon(IV) oxide, 30% in H2O, colloidal dispersion, Thermo Scientific Chemicals

產品號碼: 043111.A7
5 kg, Each
Quantity
產品號碼: 043111.A7
亦稱為 043111-A7
價格 (TWD)

化學識別

CAS
7631-86-9
IUPAC Name
silanedione
Molecular Formula
O2Si
InChI Key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES
O=[Si]=O
Concentration (weight/weight)
30% min

說明

Used as sodium stabilizing counterion, for preparation of silica catalytic supports. Also used in glass manufacturing, foundry sand casting, metallurgical flux sand, ceramics sand, gas recovery, paint and coatings, filtration sand, chromatographic column treatment, wood treatment, inks, desiccants and powder metal molding. Its is principle gateway in electronic industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

S.A. 305 m2/g
RUO – Research Use Only

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text