Thermo Scientific Chemicals

Copper(I) iodide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 043153.14
25 g, Each
Thermo Scientific Chemicals

Copper(I) iodide, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 043153.14
25 g, Each
Quantity
Catalog number: 043153.14
also known as 043153-14
Price (USD)
Price: 207.00
Online price: 175.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7681-65-4
IUPAC Name
λ¹-copper(1+) iodide
Molecular Formula
CuI
InChI Key
LSXDOTMGLUJQCM-UHFFFAOYSA-M
SMILES
[Cu+].[I-]
Total Metal Impurities
0.002% max.

Description

Copper(I) iodide is used as a catalyst in organic synthesis, cathode ray tube coating, cloud seeding, and as temperature indicator with HgI2. It catalyzes the conversion of aryl, heteroaryl, and vinyl bromides into the corresponding iodides, which are employed in coupling reactions such as Suzuki, Stille, Sonogashira, Heck and Ullmann reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text