trans-4-Octen, 97 %, Thermo Scientific Chemicals
trans-4-Octen, 97 %, Thermo Scientific Chemicals
trans-4-Octen, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

trans-4-Octen, 97 %, Thermo Scientific Chemicals

CAS: 14850-23-8 | C8H16 | 112.216 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
2 g
10 g
50 g
Katalognummer 043155.04
auch als 043155-04 bezeichnet
Preis (EUR)
49,70
Each
Menge:
2 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
49,70
Each
Chemikalien-Kennzeichnungen
CAS14850-23-8
IUPAC Name(4E)-oct-4-ene
Molecular FormulaC8H16
InChI KeyIRUCBBFNLDIMIK-BQYQJAHWSA-N
SMILESCCCC=CCCC
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Assay (GC)> 96.0%
It is used in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only