N,N'-Bis(Salicyliden)Ethylendiaminocobalt(II)-Hydrat, 96 %, Thermo Scientific Chemicals
N,N'-Bis(Salicyliden)Ethylendiaminocobalt(II)-Hydrat, 96 %, Thermo Scientific Chemicals
N,N'-Bis(Salicyliden)Ethylendiaminocobalt(II)-Hydrat, 96 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N'-Bis(Salicyliden)Ethylendiaminocobalt(II)-Hydrat, 96 %, Thermo Scientific Chemicals

CAS: 14167-18-1 | C16H14CoN2O2 | 325.23 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
1 g
5 g
25 g
100 g
Katalognummer 043174.03
auch als 043174-03 bezeichnet
Preis (EUR)
31,80
Each
Menge:
1 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
31,80
Each
Chemikalien-Kennzeichnungen
CAS14167-18-1
SpecificationsSpecification SheetDatenblatt
Appearance (Color)Orange to red to brown
Purity≥95.0%
FormPowder or crystals or crystalline powder
Salcomine catalyzes the oxidation of 2,6-disubstituted phenols by dioxygen. Experimental inhibitor of human cytμloviris proteinase activity. Metal complex dyes used as redox catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only