Thermo Scientific Chemicals

Ammonium dichromate, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 043180.22
100 g, Each
Thermo Scientific Chemicals

Ammonium dichromate, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog number: 043180.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 043180.22
also known as 043180-22
Price (USD)
Price: 561.00
Online price: 490.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7789-09-5
IUPAC Name
diammonium [(oxidodioxochromio)oxy]chromiumoylolate
Molecular Formula
Cr2H8N2O7
InChI Key
JOSWYUNQBRPBDN-UHFFFAOYSA-P
SMILES
[NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O
Appearance (Color)
Orange to red
Form
Powder or crystals or crystalline powder
Total Metal Impurities
≤0.005%

Description

Ammonium dichromate is used in photo engraving, dye mordant, pigments, pickling, photography, process engraving, lithography, pyrotechnics, glazes for porcelain and china wares. It acts as a catalyst and a source of nitrogen in the laboratory and as the photoactive site. It is also used in the preparation of alizarin and chrome alum. Further, it is used in leather and oil industry for tanning and purification respectively. It is involved in the production of the phosphor raster television screens and other devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text