Thermo Scientific Chemicals

Graphite powder, nickel coated, -100 mesh, Ni 99.8% (metals basis), Thermo Scientific Chemicals

Catalog number: 043215.36
500 g, Each
Thermo Scientific Chemicals

Graphite powder, nickel coated, -100 mesh, Ni 99.8% (metals basis), Thermo Scientific Chemicals

Catalog number: 043215.36
500 g, Each
Quantity
Catalog number: 043215.36
also known as 043215-36
Price (USD)
Form
Powder, -100 mesh
Particle Size
Report Screen Analysis, (≥90% of particles are <100 mesh)
Assay from Suppliers CofA
Nickel Content: ≥59.0%

Description

Conductive surface coatings, inks, adhesives, and sealants

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text