Thermo Scientific Chemicals

Aluminum oxide, activated, neutral, gamma-phase, 99.9% (metals basis), Thermo Scientific Chemicals

製品番号(カタログ番号): 043266.36
500 g, Each
Thermo Scientific Chemicals

Aluminum oxide, activated, neutral, gamma-phase, 99.9% (metals basis), Thermo Scientific Chemicals

製品番号(カタログ番号): 043266.36
500 g, Each
数量
製品番号(カタログ番号): 043266.36
または、製品番号043266-36

化学物質識別子

CAS
1344-28-1
IUPAC Name
dialuminium(3+) trioxidandiide
Molecular Formula
Al2O3
InChI Key
PNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Al+3].[Al+3]
Total Metal Impurities
0.1% max.

概要

Aluminum oxide is used as an adsorbent, desiccating agent and in the manufacture of dental cements and refractories. It is used in protein extraction as a component in the preparation of tissues and in bushings, spindles, lapping, polishing, refractory coating for investment casting shells, decorative non-skid tile and pressure blasting. It is also used in varnishes, for textile impregnation, in cosmetics and as an intermediate in pharmaceutical production. It is a source of aluminum in various reactions. It is used as a catalyst in a variety of reactions such as the Claus process and in the dehydration of alcohols to alkenes and as a grinding or blending agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text