Calciumsulfid, 99 % (Metallbasis ohne Mg), Mg < 1 %, Thermo Scientific Chemicals
Calciumsulfid, 99 % (Metallbasis ohne Mg), Mg &lt; 1 %, Thermo Scientific Chemicals
Calciumsulfid, 99 % (Metallbasis ohne Mg), Mg &lt; 1 %, Thermo Scientific Chemicals
Calciumsulfid, 99 % (Metallbasis ohne Mg), Mg &lt; 1 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calciumsulfid, 99 % (Metallbasis ohne Mg), Mg < 1 %, Thermo Scientific Chemicals

CAS: 20548-54-3 | CaS | 72.14 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
25 g
100 g
Katalognummer 043646.14
auch als 043646-14 bezeichnet
Preis (EUR)
257,00
Each
Zum Warenkorb hinzufügen
Menge:
25 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
257,00
Each
Zum Warenkorb hinzufügen
Chemikalien-Kennzeichnungen
CAS20548-54-3
IUPAC Namecalcium sulfanediide
Molecular FormulaCaS
InChI KeyAGVJBLHVMNHENQ-UHFFFAOYSA-N
SMILES[S--].[Ca++]
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Impurity contentMg <1%
Total Metal Impurities1% max.
Calcium sulfide is used in phosphors, lubricant additives and luminous paints. It reacts with elemental sulfur to give a lime-sulfur. Further, it is used in ore dressing and flotation agent. In addition to this, it is used as a depilatory in the tanning industry and in cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only