Thermo Scientific Chemicals

Palladium(II) bromide, Premion™, 99.998% (metals basis), Pd 39.5% min, Thermo Scientific Chemicals

Catalog number: 043697.06
5 g, Each
Thermo Scientific Chemicals

Palladium(II) bromide, Premion™, 99.998% (metals basis), Pd 39.5% min, Thermo Scientific Chemicals

Catalog number: 043697.06
5 g, Each
Quantity
Catalog number: 043697.06
also known as 043697-06
Price (USD)

Chemical Identifiers

CAS
13444-94-5
IUPAC Name
palladium(2+) dibromide
Molecular Formula
Br2Pd
InChI Key
INIOZDBICVTGEO-UHFFFAOYSA-L
SMILES
[Br-].[Br-].[Pd++]
Total Metal Impurities
0.002% max.

Description

Palladium(II) bromide is used as a source of the palladium(0) precatalyst in various coupling reactions. It is used as a catalyst for the carbonylative Heck coupling reaction. It is also used as an oxidizing agent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text