Tantal(V)-chlorid, wasserfrei, Puratronic™, 99.995 % (Metallbasis), Thermo Scientific Chemicals
Tantal(V)-chlorid, wasserfrei, Puratronic™, 99.995 % (Metallbasis), Thermo Scientific Chemicals
Tantal(V)-chlorid, wasserfrei, Puratronic™, 99.995 % (Metallbasis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tantal(V)-chlorid, wasserfrei, Puratronic™, 99.995 % (Metallbasis), Thermo Scientific Chemicals

CAS: 7721-01-9 | Cl5Ta | 358.20 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
5 g
25 g
100 g
Katalognummer 043731.14
auch als 043731-14 bezeichnet
Preis (EUR)
210,00
Each
Menge:
25 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
210,00
Each
Chemikalien-Kennzeichnungen
CAS7721-01-9
IUPAC Nametantalum(5+) pentachloride
Molecular FormulaCl5Ta
InChI KeyOEIMLTQPLAGXMX-UHFFFAOYSA-I
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ta+5]
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Total Metal Impurities0.005% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampulliert unter Argon
RUO – Research Use Only