Thermo Scientific Chemicals

Polyethylene, low density, 1000 micron, Thermo Scientific Chemicals

Catalog number: 043949.30
250 g, Each
Thermo Scientific Chemicals

Polyethylene, low density, 1000 micron, Thermo Scientific Chemicals

Catalog number: 043949.30
250 g, Each
Quantity
Catalog number: 043949.30
also known as 043949-30
Price (USD)
Price: 55.30
Online price: 47.65
Your price:
Quantity
-

Chemical Identifiers

CAS
9002-88-4
Molecular Formula
(C2H4)n
SMILES
*-CC-*
MDL Number
MFCD00084423
Comment
Density 0.92 to 0.94 g/ml
Appearance (Color)
White
Form
Granule

Description

Polyethylene is used as an additive to unsaturated polyesters, epoxides and other polymers to impart the unique properties of ultra-high-molecular-weight polyethylene (UHMWPE). It is widely used for the manufacturing of various containers, trays, dispensing bottles, plastic wraps, plastic bags for computer components and various molded laboratory equipments. It is used in the manufacture of plastic bags, yard bags or outdoor bags, which is an environmentally sensible material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text