Thermo Scientific Chemicals

Bismuth(III) telluride, 99.98% (metals basis), Thermo Scientific Chemicals

Catalog number: 044077.14
25 g, Each
Thermo Scientific Chemicals

Bismuth(III) telluride, 99.98% (metals basis), Thermo Scientific Chemicals

Catalog number: 044077.14
25 g, Each
Quantity
Catalog number: 044077.14
also known as 044077-14
Price (USD)
Price: 253.00
Online price: 218.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1304-82-1
IUPAC Name
dibismuth(3+) tritellanediide
Molecular Formula
Bi2Te3
InChI Key
AZFMNKUWQAGOBM-UHFFFAOYSA-N
SMILES
[Te--].[Te--].[Te--].[Bi+3].[Bi+3]
Total Metal Impurities
0.02% max.

Description

Bismuth(III) telluride and its alloys (antimony or selenium) posses excellent semiconductor activity, which is used in thermoelectric industry for cooling and energy generation. It increases the speed of microchips and acts as topological insulators in spintronics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text