Fulleren, Nanoröhrchen, mehrwandig, 20–50 nm AD, 5–20 Mikrometer lang, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Fulleren, Nanoröhrchen, mehrwandig, 20–50 nm AD, 5–20 Mikrometer lang, Thermo Scientific Chemicals

CAS: 308068-56-6 | CH4 | 16.043 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
1 g
5 g
Katalognummer 044192.06
auch als 044192-06 bezeichnet
Preis (EUR)
1.006,00
Each
Menge:
5 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
1.006,00
Each
Chemikalien-Kennzeichnungen
CAS308068-56-6
SpecificationsSpecification SheetDatenblatt
FormPowder
Assay from Supplier's CofA≥94.0%
DiameterReport (nm)
LengthReport (nm)
Carbon nanotubes enhance the electrical conductivity and the mechanical properties. It is also used in chemical research. It is suitable as a hydrogen storage material. It is an ideal super capacitor electrode materials, It is used in the preparation of super composites. carbon nanotubes have excellent field emission performance and used in heavy weight cathode tube technology. It is also applied to naanometer electronic, catalytic fiber machine. Nanotubes will have an important role in nanotechnology engineering. carbon nanotube also can be used in making molecule bearings and nano robot.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only