Stahl-Shot, nicht kugelförmig, 1 mm (0.039 Zoll) Durchmesser, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Stahl-Shot, nicht kugelförmig, 1 mm (0.039 Zoll) Durchmesser, Thermo Scientific Chemicals

Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
2 kg
5 kg
Katalognummer 044236.A3
auch als 044236-A3 bezeichnet
Preis (EUR)
-
Menge:
2 kg
Großbestellung oder individuelle Größe anfordern
Specifications
Chemischer Name oder MaterialSteel shot
MDL-NummerMFCD00678174
Namenshinweis1mm (0.039 in.) dia.
Empfohlene LagerungUmgebungstemperaturen
LöslichkeitsinformationenIt is insoluble in water.
Mehr anzeigen
Steel shot is used in cleaning applications for removal of loose material on metal surfaces. This type of cleaning is common in automotive industry (motor blocks, cylinder heads. Steel shot is also used in surface preparation process for cleaning metal surfaces which are covered with mill scale, dirt, rust, or paint coatings and for physically modifying the metal surface such as creating roughness for better application of paint and coating. The steel shots are generally employed in shot blasting machines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only