Cer(III)-sulfat-Oktahydrat, REacton™, 99.999 % (Metallbasis), Thermo Scientific Chemicals
Cer(III)-sulfat-Oktahydrat, REacton™, 99.999 % (Metallbasis), Thermo Scientific Chemicals
Cer(III)-sulfat-Oktahydrat, REacton™, 99.999 % (Metallbasis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cer(III)-sulfat-Oktahydrat, REacton™, 99.999 % (Metallbasis), Thermo Scientific Chemicals

CAS: 10450-59-6 | Ce2(SO4)3·8H2O
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
50 g
250 g
Katalognummer 044357.30
auch als 044357-30 bezeichnet
Preis (EUR)
802,00
Each
Menge:
250 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
802,00
Each
Chemikalien-Kennzeichnungen
CAS10450-59-6
IUPAC Namedicerium(3+) octahydrate trisulfate
Molecular FormulaCe2H16O20S3
InChI KeyPQBKXYUMEMUVIH-UHFFFAOYSA-H
SMILESO.O.O.O.O.O.O.O.[Ce+3].[Ce+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Total Metal Impurities0.001% max.
Cerium(III) sulfate octahydrate used in waterproofing, mildew-proofing, and in dyeing and printing textiles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only