Thermo Scientific Chemicals

3-Hydroxy-5-methylisoxazole, 97%, Thermo Scientific Chemicals

Catalog number: 044450.22
100 g, Each
Thermo Scientific Chemicals

3-Hydroxy-5-methylisoxazole, 97%, Thermo Scientific Chemicals

Catalog number: 044450.22
100 g, Each
Quantity
Catalog number: 044450.22
Price (USD)
Price: 518.00
Online price: 451.65
Your price:
Quantity
-

Chemical Identifiers

CAS
10004-44-1
IUPAC Name
5-methyl-2,3-dihydro-1,2-oxazol-3-one
Molecular Formula
C4H5NO2
InChI Key
KGVPNLBXJKTABS-UHFFFAOYSA-N
SMILES
CC1=CC(=O)NO1
Appearance (Color)
Off-white to yellow
Form
powder
Assay (GC)
> 96.0%

Description

3-Hydroxy-5-methylisoxazole is used in plant growth stimulation activity and seedlings growing. And it is also used in DNA/RNA synthesis inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text