n-Heptan, wasserfrei, ≥99 %, verpackt unter Argon in wiederverschließbaren ChemSeal™ Flaschen, Thermo Scientific Chemicals
n-Heptan, wasserfrei, ≥99 %, verpackt unter Argon in wiederverschließbaren ChemSeal™ Flaschen, Thermo Scientific Chemicals
n-Heptan, wasserfrei, ≥99 %, verpackt unter Argon in wiederverschließbaren ChemSeal™ Flaschen, Thermo Scientific Chemicals
Thermo Scientific Chemicals

n-Heptan, wasserfrei, ≥99 %, verpackt unter Argon in wiederverschließbaren ChemSeal™ Flaschen, Thermo Scientific Chemicals

CAS: 142-82-5 | C7H16 | 100.21 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
100 mL
1 L
Katalognummer 044457.K2
auch als 044457-K2 bezeichnet
Preis (EUR)
78,65
Exklusiv online
86,80
Ersparnis 8,15 (9%)
Each
Menge:
1 L
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
78,65
Exklusiv online
86,80
Ersparnis 8,15 (9%)
Each
Chemikalien-Kennzeichnungen
CAS142-82-5
IUPAC Nameheptane
Molecular FormulaC7H16
InChI KeyIMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILESCCCCCCC
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)Colorless
Assay (GC)≥99%
CommentInert gas used can be either Argon or Nitrogen
Water Content (Karl Fischer Titration)≤10 ppm
FormLiquid
Mehr anzeigen
n-Heptane is used in paints and coatings, as the rubber cement solvent and GC analysis, HPLC, spectrophotometry and environmental testing. Used as a primary reference standard in determining the knock rating of carburetor fuels and solvent especially for vegetable oils, low temperature thermo- meters, calibrations, and polymerization reaction medium. The hydroisomerization of n-heptane to form dibranched and tribranched products useful for producing high octane gasoline. water/AOT/n-heptane microemulsions plays an essential role for the synthesis, size control, and passivation of CdS nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Verpackt unter Argon
RUO – Research Use Only