Polyphenylmethylsiloxan, Molekulargewicht 2500 bis 2700, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Polyphenylmethylsiloxan, Molekulargewicht 2500 bis 2700, Thermo Scientific Chemicals

CAS: 9005-12-3 | C7H8OSi | 136.23 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
50 g
250 g
1 kg
Katalognummer 044569.A1
auch als 044569-A1 bezeichnet
Preis (EUR)
519,00
Each
Menge:
1 kg
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
519,00
Each
Chemikalien-Kennzeichnungen
CAS9005-12-3
IUPAC Namemethyl(phenyl)silanone
Molecular FormulaC7H8OSi
InChI KeyIXKNJYSAWNJQQY-UHFFFAOYSA-N
SMILESC[Si](=O)C1=CC=CC=C1
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)Clear, colorless
FormViscous liquid
Viscosity450-550 cSt
Infrared spectrumConforms to structure
A biochemical for proteomics research. A novel coating for solid-phase microextraction. A polyphenylmethylsiloxane for use in skincare, suncare, color cosmetic, hair care, and antiperspirant / deodorant applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only