(S)-(-)-2,2',6,6'-Tetramethoxy-4,4'-bis-(di-(3,5-xylyl)-phosphino)-3,3'-bipyridin, Thermo Scientific Chemicals
(S)-(-)-2,2',6,6'-Tetramethoxy-4,4'-bis-(di-(3,5-xylyl)-phosphino)-3,3'-bipyridin, Thermo Scientific Chemicals
(S)-(-)-2,2',6,6'-Tetramethoxy-4,4'-bis-(di-(3,5-xylyl)-phosphino)-3,3'-bipyridin, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(S)-(-)-2,2',6,6'-Tetramethoxy-4,4'-bis-(di-(3,5-xylyl)-phosphino)-3,3'-bipyridin, Thermo Scientific Chemicals

Asymmetrische Hydrierung
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
0.1 g
0.5 g
Katalognummer 044614.02
auch als 044614-02 bezeichnet
Preis (EUR)
293,00
Each
Menge:
0.5 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
293,00
Each
Specifications
CAS443347-10-2
Chemischer Name oder Material(S)- (-)-2,2',6,6'-Tetramethoxy-4,4'-bis (di (3,5-xylyl)phosphino)-3,3'-bipyridine
Formelmasse756.86
IUPAC-Name[3-[4-Bis(3,5-Dimethylphenyl)Phosphanyl-2,6-Dimethoxypyridin-3-yl]-2,6-Dimethoxypyridin-4-yl]-Bis(3,5-Dimethylphenyl)Phosphan
InChI-SchlüsselJRTHAKOHBMETRC-UHFFFAOYSA-N
Mehr anzeigen
(S)-(-)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridine is used in Pharmaceutical and industry applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Verpackt unter Argon
RUO – Research Use Only