Nickel(II)-oxid, grün, Ni 78.5 %, Thermo Scientific Chemicals
Nickel(II)-oxid, grün, Ni 78.5 %, Thermo Scientific Chemicals
Nickel(II)-oxid, grün, Ni 78.5 %, Thermo Scientific Chemicals
Nickel(II)-oxid, grün, Ni 78.5 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel(II)-oxid, grün, Ni 78.5 %, Thermo Scientific Chemicals

CAS: 1313-99-1 | NiO | 74.69 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
250 g
1 kg
Katalognummer 045094.A1
auch als 045094-A1 bezeichnet
Preis (EUR)
339,00
Each
Zum Warenkorb hinzufügen
Menge:
1 kg
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
339,00
Each
Zum Warenkorb hinzufügen
Chemikalien-Kennzeichnungen
CAS1313-99-1
IUPAC Nameoxonickel
Molecular FormulaNiO
InChI KeyGNRSAWUEBMWBQH-UHFFFAOYSA-N
SMILESO=[Ni]
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)Green
FormPowder
Particle Size-400 Mesh
Assay (special)≥78.5% Ni
Nickel(II) oxide is used in the preparation of nickel alloys including nickel steel alloys, manufacture of glass and porcelain paints. Further nickel oxide is the main component in nickel-iron battery (Edison battery), nickel-cadmium rechargeable battery, and also used in ceramic industry for making frits and porcelain glazes. It also acts as a hydrogenation catalyst. NiO/CNTs (nickel oxide/carbon nanotubes) could be a potential cathode catalyst for oxygen reduction reaction (ORR) in microbial fuel cells (MFCs). It is blended with other high purity oxides and used in a variety of semiconductor applications such as thermistors and varistors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only