Arsen(III)-sulfid, 99.999 % (Metallbasis), Thermo Scientific Chemicals
Arsen(III)-sulfid, 99.999 % (Metallbasis), Thermo Scientific Chemicals
Arsen(III)-sulfid, 99.999 % (Metallbasis), Thermo Scientific Chemicals
Arsen(III)-sulfid, 99.999 % (Metallbasis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Arsen(III)-sulfid, 99.999 % (Metallbasis), Thermo Scientific Chemicals

CAS: 1303-33-9 | As2S3 | 246.02 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
5 g
25 g
Katalognummer 045522.14
auch als 045522-14 bezeichnet
Preis (EUR)
558,00
Each
Zum Warenkorb hinzufügen
Menge:
25 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
558,00
Each
Zum Warenkorb hinzufügen
Chemikalien-Kennzeichnungen
CAS1303-33-9
IUPAC Namediarsenic(3+) trisulfanediide
Molecular FormulaAs2S3
InChI KeyCUGMJFZCCDSABL-UHFFFAOYSA-N
SMILES[S--].[S--].[S--].[As+3].[As+3]
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Total Metal Impurities0.001% max.
Arsenic(III) sulfide is used to make infrared-transmitting glass, pyrotechnics, pigments, linoleum, electrical semiconductors and photoconductors. It acts as a reducing agent and serves as a tanning agent. It is also used for depilating hides and veterinary medication. Further, it is used as an inorganic photoresist to fabricate 3-D nanostructures. In addition to this, it is used in the treatment for acute promyelocytic leukemia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only