Aluminiumoxid, superaktiviert, sauer, Gütegrad I, Thermo Scientific Chemicals
Aluminiumoxid, superaktiviert, sauer, Gütegrad I, Thermo Scientific Chemicals
Aluminiumoxid, superaktiviert, sauer, Gütegrad I, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminiumoxid, superaktiviert, sauer, Gütegrad I, Thermo Scientific Chemicals

CAS: 1344-28-1 | Al2O3 | 101.96 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
250 g
1 kg
Katalognummer 046065.A1
auch als 046065-A1 bezeichnet
Preis (EUR)
-
Menge:
1 kg
Großbestellung oder individuelle Größe anfordern
Chemikalien-Kennzeichnungen
CAS1344-28-1
IUPAC Namedialuminium(3+) trioxidandiide
Molecular FormulaAl2O3
InChI KeyPNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Al+3].[Al+3]
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Assay (special)Ignition LOss: ≤3.0%
pH4.0-5.5
Particle SizeParticle Distribution (50-200 micron): ≥95%
Particle SizePore Volume: Report
Appearance (Color)White
Mehr anzeigen
Aluminum oxide is an inorganic compound used as a grinding or blending agent. In addition, Aluminum oxide can be essential in protein extraction as a component in the preparation of tissues. Aluminum oxide is a source of aluminum in reactions, an abrasive agent, and as a refractory material. Also Aluminum oxide can act as a catalyst in a variety of reactions such as the Claus process and in the dehydration of alcohols to alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only