Bismuth(III)-oxid, Nanopulver, 99.9 %, Thermo Scientific Chemicals
Bismuth(III)-oxid, Nanopulver, 99.9 %, Thermo Scientific Chemicals
Bismuth(III)-oxid, Nanopulver, 99.9 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bismuth(III)-oxid, Nanopulver, 99.9 %, Thermo Scientific Chemicals

CAS: 1304-76-3 | Bi2O3 | 465.96 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
25 g
100 g
500 g
Katalognummer 046314.22
auch als 046314-22 bezeichnet
Preis (EUR)
106,00
Each
Menge:
100 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
106,00
Each
Chemikalien-Kennzeichnungen
CAS1304-76-3
IUPAC Namedibismuth(3+) trioxidandiide
Molecular FormulaBi2O3
InChI KeyTYIXMATWDRGMPF-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Bi+3].[Bi+3]
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)Yellow
Assay (special)≥99.9% Bi
FormPowder
Particle Size80-200nm Average
Bismuth(III) oxide is used in the preparation of BiFeO3perovskite nanoparticles. It finds use in disinfectants, magnets, glass, rubber, vulcanization, fireproofing papers and polymers and catalysts. Bismuth trioxide brings about the dragon's eggs effect in fireworks, as a replacement of red lead. Bismuth(III) compounds are attractive reagents and catalysts in organic synthesis because of their low cost and ease of handling. Bismuth trioxide nanoparticles also play an important role in high energy gas generators. The alpha crystalline form of bismuth(III) oxide has p-type electronic conductivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only