Thermo Scientific Chemicals

Rhodium(III) chloride solution, 99.5% (metals basis), Rh 10-30% w/w (cont. Rh), Thermo Scientific Chemicals

Catalog number: 046523.NJ
0.5 g, Each
Thermo Scientific Chemicals

Rhodium(III) chloride solution, 99.5% (metals basis), Rh 10-30% w/w (cont. Rh), Thermo Scientific Chemicals

Catalog number: 046523.NJ
0.5 g, Each
Quantity
Catalog number: 046523.NJ
also known as 046523-NJ
Price (USD)

Chemical Identifiers

CAS
10049-07-7
IUPAC Name
rhodium(3+) trichloride
Molecular Formula
Cl3Rh
InChI Key
SONJTKJMTWTJCT-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Rh+3]
Assay (unspecified)
Rh 10-30%
Total Metal Impurities
0.5% max.
Form
Liquid

Description

It is used as primary and secondary intermediate, in the preparation of compounds used in homogeneous catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text