Thermo Scientific Chemicals

Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II), Thermo Scientific Chemicals

Catalog number: 046803.MD
250 mg, Each
Thermo Scientific Chemicals

Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II), Thermo Scientific Chemicals

Catalog number: 046803.MD
250 mg, Each
Quantity
Catalog number: 046803.MD
also known as 046803-MD
Price (USD)
Price: 134.00
Online price: 113.65
Your price:
Quantity
-

Chemical Identifiers

CAS
92390-26-6
IUPAC Name
Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II)
Molecular Formula
C18H27ClRu
SMILES
Cl[Ru++].C1C\C=C/CC\C=C/1.Cc1c(C)c(C)c(C)c1C
Molecular Weight (g/mol)
379.93
Appearance (Color)
Yellow to brown
Form
Powder
Assay (unspecified)
55.90 - 57.90% C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text