Gold sputtering target, 57.0mm (2.2in) dia x 0.3mm (0.012in) thick, 99.99% (metals basis)
Thermo Scientific Chemicals

Gold sputtering target, 57.0mm (2.2in) dia x 0.3mm (0.012in) thick, 99.99% (metals basis)

CAS: 7440-57-5
Have Questions?
KatalognummerMenge
047091.KS
auch als 047091-KS bezeichnet
1 Ea.
Katalognummer 047091.KS
auch als 047091-KS bezeichnet
Preis (EUR)
-
Menge:
1 Ea.
Großbestellung oder individuelle Größe anfordern
Chemikalien-Kennzeichnungen
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
Appearance (Color)Gold
FormDisc
Comment0.27 - 0.33 mm thick
Diameter51.3 - 62.7mm
Total Metal Impurities0.01% max.
Gold sputtering target metal is retained for use as bullion reserves, but some is used within the electronics and jewellery industries, where it is frequently alloyed with other elements to improve the mechanical properties of the metal (e.g. copper and silver). Other uses for the metal are as a heat reflecting coating for glass as well as as a decorative medium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.