Zinn(IV)-iodid, 95 %, Thermo Scientific Chemicals
Zinn(IV)-iodid, 95 %, Thermo Scientific Chemicals
Zinn(IV)-iodid, 95 %, Thermo Scientific Chemicals
Zinn(IV)-iodid, 95 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinn(IV)-iodid, 95 %, Thermo Scientific Chemicals

CAS: 7790-47-8 | H4I4Sn | 630.36 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
10 g
50 g
Katalognummer 071114.18
auch als 071114-18 bezeichnet
Preis (EUR)
269,00
Each
Zum Warenkorb hinzufügen
Menge:
50 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
269,00
Each
Zum Warenkorb hinzufügen
Chemikalien-Kennzeichnungen
CAS7790-47-8
IUPAC Nametin(IV) iodide
Molecular FormulaH4I4Sn
InChI KeyZBCVLXKKLMNLGV-UHFFFAOYSA-N
SMILES[Sn].I.I.I.I
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥94.0%
Appearance (Color)Yellow to orange
Used in chemical research and as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only