Thermo Scientific Chemicals

Strontium bromide hexahydrate, 95%, Thermo Scientific Chemicals

Catalog number: 072105.36
500 g, Each
Thermo Scientific Chemicals

Strontium bromide hexahydrate, 95%, Thermo Scientific Chemicals

Catalog number: 072105.36
500 g, Each
Quantity
Catalog number: 072105.36
also known as 072105-36
Price (USD)
Price: 127.00
Online price: 107.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7789-53-9
IUPAC Name
strontium(2+) hexahydrate dibromide
Molecular Formula
Br2H12O6Sr
InChI Key
FLMJUJXBFKFYOZ-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Br-].[Br-].[Sr++]
Assay (unspecified)
94.1% min

Description

Strontium bromide hexahydrate is used in flares and also used in some pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text